Methyl (4-bromo-3-fluorophenyl)acetate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(4-bromo-3-fluorophenyl)acetate

114642-1G
102894-414EA 958.46 USD
102894-414
Methyl (4-bromo-3-fluorophenyl)acetate ≥97%
Methyl (4-bromo-3-fluorophenyl)acetate

Hazard: Irritant

Formula: C₉H₈BrFO₂
MW: 247.07 g/mol
MDL Number: MFCD22418249
CAS Number: 942282-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR