4-Methylisoxazole-5-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114552-1G
102894-282EA 958.46 USD
102894-282
4-Methylisoxazole-5-carbonyl chloride ≥97%
4-Methylisoxazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₅H₄ClNO₂
MDL Number: MFCD28099738
CAS Number: 1108713-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR