4-Bromo-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

114529-1G
102894-232EA 1104.62 USD
102894-232
4-Bromo-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ≥97%
4-Bromo-3,5-dimethyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₈BrNO₂
MDL Number: MFCD11557240
CAS Number: 562074-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR