1-Bromo-2-methyl-3,5-bis(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

114058-1G
102893-228EA 813.85 USD
102893-228
1-Bromo-2-methyl-3,5-bis(trifluoromethyl)benzene ≥95%
1-Bromo-2-methyl-3,5-bis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₉H₅BrF₆
MDL Number: MFCD01631703
CAS Number: 251651-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR