3-(Anilinomethyl)-7-methylquinolin-2(1H)-one ≥95%

Supplier: Matrix Scientific

115166-500MG 115166-1G
102892-194EA 693.85 USD
102892-194 102892-196
3-(Anilinomethyl)-7-methylquinolin-2(1H)-one ≥95%
3-(Anilinomethyl)-7-methylquinolin-2(1H)-one

Hazard: Irritant

Formula: C₁₇H₁₆N₂O
MW: 264.33 g/mol
MDL Number: MFCD04072102
CAS Number: 573696-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR