(2R,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate hydrochloride

Supplier: Matrix Scientific

Synonyms: rel-(2R,5S)-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate hydrochloride

113362-1G
102891-978EA 1832.31 USD
102891-978
(2R,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate hydrochloride
(2R,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O₂
MW: 250.77 g/mol
MDL Number: MFCD13191714
CAS Number: 792969-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR