6-Bromo-5-fluoro-1H-indazole

Supplier: Matrix Scientific

113199-1G
102891-644EA 1018.46 USD
102891-644
6-Bromo-5-fluoro-1H-indazole
6-Bromo-5-fluoro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrFN₂
MDL Number: MFCD18089816
CAS Number: 1286734-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR