Human Recombinant Snap Alpha
Supplier: Bon Opus Biosciences
C275-10UG C275-1MG C275-500UG C275-50UG
102861-006EA 170.15 USD
102861-006 102861-008 102861-010 102861-012
Human Recombinant Snap Alpha
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human SNAP-alpha is produced by our E.coli expression system and the target gene encoding Met1-Arg295 is expressed with a 6His tag at the N-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx.
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.