Human Recombinant C-X-C Motif Chemokine 2
Supplier: Bon Opus Biosciences
C096-10UG C096-1MG C096-500UG C096-50UG
102859-590EA 170.15 USD
102859-590 102859-592 102859-594 102859-596
Human Recombinant C-X-C Motif Chemokine 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human C-X-C Motif Chemokine 2 is produced by our E.coli expression system and the target gene encoding Thr39-Asn107 is expressed.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.