Human Recombinant Leukemia Inhibitory Factor

Supplier: Bon Opus Biosciences

C017-10UG C017-1MG C017-500UG C017-50UG
102859-094EA 118.46 USD
102859-094 102859-096 102859-098 102859-100
Human Recombinant Leukemia Inhibitory Factor
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human Leukemia Inhibitory Factor is produced by our E.coli expression system and the target gene encoding Ser23-Phe202 is expressed.


Información accesorios: Accessories not included.


Información de suministro: Shipped Via FedEx


Envase: Shipped at ambient temperature via FedEx envelope.


Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now

SPECIFICATIONS

Protein/Peptide Name Leukemia Inhibitory Factor
Environmentally Preferable
Protein/Peptide Type Recombinant
Species Human
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR