2-Naphthyl trifluoromethyl ketone ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(naphthalen-2-yl)ethanone

086919-1G 086919-2G 086919-5G
102851-416EA 520 USD
102851-416 102851-418 102851-420
2-Naphthyl trifluoromethyl ketone ≥97%
2-Naphthyl trifluoromethyl ketone

Hazard: Irritant

Formula: C₁₂H₇F₃O
MW: 224.18 g/mol
Melting Pt: 40…41 °C
MDL Number: MFCD01320008
CAS Number: 1800-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR