3-(4-n-Pentylphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086248-2G
102847-568EA 560 USD
102847-568
3-(4-n-Pentylphenyl)-1-propene ≥97%
3-(4-n-Pentylphenyl)-1-propene

Hazard: Irritant

Formula: C₁₄H₂₀
MDL Number: MFCD09801163
CAS Number: 951890-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR