4-Hydrazino-7-methylthieno[3,2-d]pyrimidine ≥97%

Supplier: Matrix Scientific

085947-1G
102846-454EA 496.92 USD
102846-454
4-Hydrazino-7-methylthieno[3,2-d]pyrimidine ≥97%
4-Hydrazino-7-methylthieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₇H₈N₄S
MDL Number: MFCD00205196
CAS Number: 175137-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR