6,7,8,9-Tetrahydro-5H-imidazo[1,2-a]azepine ≥97%

Supplier: Matrix Scientific

085921-1G
102846-388EA 750.77 USD
102846-388
6,7,8,9-Tetrahydro-5H-imidazo[1,2-a]azepine ≥97%
6,7,8,9-Tetrahydro-5H-imidazo[1,2-a]azepine

Hazard: Irritant

Formula: C₈H₁₂N₂
MDL Number: MFCD11870772
CAS Number: 5768-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR