1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol ≥97%

Supplier: Matrix Scientific

085783-1G
102845-978EA 636.92 USD
102845-978
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol ≥97%
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol

Hazard: Irritant

Formula: C₁₄H₁₃FO
MW: 216.26 g/mol
MDL Number: MFCD18089274
CAS Number: 56430-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR