3-(4-Methylpiperazin-1-yl)-6-(trifluoromethyl)pyridazine ≥97%

Supplier: Matrix Scientific

085472-1G
102845-124EA 806.15 USD
102845-124
3-(4-Methylpiperazin-1-yl)-6-(trifluoromethyl)pyridazine ≥97%
3-(4-Methylpiperazin-1-yl)-6-(trifluoromethyl)pyridazine

Hazard: Irritant

Formula: C₁₀H₁₃F₃N₄
MW: 246.24 g/mol
MDL Number: MFCD11840307
CAS Number: 1217006-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR