2-Methyl-4-(methylthio)-6-oxo-1,6-dihydropyrimidine-5-carbonitrile ≥97%

Supplier: Matrix Scientific

085458-1G
102845-094EA 723.08 USD
102845-094
2-Methyl-4-(methylthio)-6-oxo-1,6-dihydropyrimidine-5-carbonitrile ≥97%
2-Methyl-4-(methylthio)-6-oxo-1,6-dihydropyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₇H₇N₃OS
MDL Number: MFCD00067845
CAS Number: 15908-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR