1-[4-(Trifluoromethyl)phenyl]cyclobutanecarbonitrile ≥97%

Supplier: Matrix Scientific

084865-1G
102843-598EA 750.77 USD
102843-598
1-[4-(Trifluoromethyl)phenyl]cyclobutanecarbonitrile ≥97%
1-[4-(Trifluoromethyl)phenyl]cyclobutanecarbonitrile

Hazard: Irritant

Formula: C₁₂H₁₀F₃N
MDL Number: MFCD11036721
CAS Number: 29786-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR