2-Methyl-2-(4-nitrophenyl)propanenitrile ≥97%

Supplier: Matrix Scientific

084797-1G
102843-402EA 636.92 USD
102843-402
2-Methyl-2-(4-nitrophenyl)propanenitrile ≥97%
2-Methyl-2-(4-nitrophenyl)propanenitrile

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD00844930
CAS Number: 71825-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR