6-(Hydroxymethyl)-2-naphthol ≥97%

Supplier: Matrix Scientific

Synonyms: 6-(Hydroxymethyl)naphthalen-2-ol

084293-1G
102841-996EA 1118.46 USD
102841-996
6-(Hydroxymethyl)-2-naphthol ≥97%
6-(Hydroxymethyl)-2-naphthol

Hazard: Irritant

Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
MDL Number: MFCD12756488
CAS Number: 309752-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR