4-(Hydroxymethyl)cyclohexanone ≥97%

Supplier: Matrix Scientific

084270-1G
102841-938EA 200 USD
102841-938
4-(Hydroxymethyl)cyclohexanone ≥97%
4-(Hydroxymethyl)cyclohexanone

Hazard: Irritant

Formula: C₇H₁₂O₂
MW: 128.1690 g/mol
Boiling Pt: 105…110 °C (0.08 mmHg)
MDL Number: MFCD08056334
CAS Number: 38580-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR