5-Bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole ≥97%

Supplier: Matrix Scientific

083913-1G
102841-056EA 170.77 USD
102841-056
5-Bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole ≥97%
5-Bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole

Hazard: Irritant

Formula: C₁₁H₇BrF₃NS
MW: 322.15 g/mol
MDL Number: MFCD04974053
CAS Number: 850375-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR