2-Iodo-4-methylpyridine ≥97%

Supplier: Matrix Scientific

083867-1G
102840-934EA 636.92 USD
102840-934
2-Iodo-4-methylpyridine ≥97%
2-Iodo-4-methylpyridine

Hazard: Irritant

Formula: C₆H₆IN
MDL Number: MFCD12963767
CAS Number: 22282-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR