5-(3-Chlorophenyl)oxazole ≥97%

Supplier: Matrix Scientific

083783-1G 083783-10G
102840-716EA 255.38 USD
102840-716 102840-718
5-(3-Chlorophenyl)oxazole ≥97%
5-(3-Chlorophenyl)oxazole

Hazard: Irritant

Formula: C₉H₆ClNO
MW: 179.6000 g/mol
MDL Number: MFCD00084974
CAS Number: 89808-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR