Ethyl quinoxaline-6-carboxylate ≥97%

Supplier: Matrix Scientific

083551-1G
102840-066EA 298.46 USD
102840-066
Ethyl quinoxaline-6-carboxylate ≥97%
Ethyl quinoxaline-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
MDL Number: MFCD09998746
CAS Number: 6924-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR