3-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride ≥95%

Supplier: Matrix Scientific

083108-1G
102838-936EA 806.15 USD
102838-936
3-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride ≥95%
3-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride

Hazard: Irritant

Formula: C₁₁H₈ClNOS
MDL Number: MFCD06659063
CAS Number: 844891-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR