4-Vinylbenzoyl chloride ≥95%

Supplier: Matrix Scientific

083092-1G
102838-896EA 806.15 USD
102838-896
4-Vinylbenzoyl chloride ≥95%
4-Vinylbenzoyl chloride

Hazard: Irritant

Formula: C₉H₇ClO
MW: 166.61 g/mol
MDL Number: MFCD13173795
CAS Number: 1565-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR