1-Benzyl-4-hydroxypiperidine-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082758-1G
102838-056EA 949.23 USD
102838-056
1-Benzyl-4-hydroxypiperidine-4-carboxylic acid ≥97%
1-Benzyl-4-hydroxypiperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MW: 235.29 g/mol
MDL Number: MFCD06208305
CAS Number: 59119-18-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR