2-[(3-Nitro-2-pyridyl)thio]acetic acid ≥97%

Supplier: Matrix Scientific

082387-1G
102837-234EA 467.69 USD
102837-234
2-[(3-Nitro-2-pyridyl)thio]acetic acid ≥97%
2-[(3-Nitro-2-pyridyl)thio]acetic acid

Hazard: Irritant

Formula: C₇H₆N₂O₄S
MW: 214.2 g/mol
MDL Number: MFCD00174245
CAS Number: 18504-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR