N-Methyl-N-[(2-methyl-1H-indol-3-yl)methyl]amine ≥97%

Supplier: Matrix Scientific

081938-1G
102836-156EA 566.15 USD
102836-156
N-Methyl-N-[(2-methyl-1H-indol-3-yl)methyl]amine ≥97%
N-Methyl-N-[(2-methyl-1H-indol-3-yl)methyl]amine

Hazard: Irritant

Formula: C₁₁H₁₄N₂
MDL Number: MFCD08056294
CAS Number: 335032-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR