1-[6-(Trifluoromethyl)pyridazin-3-yl]piperidin-4-amine ≥97%

Supplier: Matrix Scientific

081401-1G
102834-810EA 863.08 USD
102834-810
1-[6-(Trifluoromethyl)pyridazin-3-yl]piperidin-4-amine ≥97%
1-[6-(Trifluoromethyl)pyridazin-3-yl]piperidin-4-amine

Hazard: Irritant

Formula: C₁₀H₁₃F₃N₄
MDL Number: MFCD11840262
CAS Number: 1211538-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR