2-Bromo-4'-(dimethylamino)acetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Dimethylamino)phenacyl bromide, 2-Bromo-1-(4-(dimethylamino)phenyl)ethanone

081081-1G
102833-992EA 750.77 USD
102833-992
2-Bromo-4'-(dimethylamino)acetophenone ≥97%
2-Bromo-4'-(dimethylamino)acetophenone

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MW: 242.1124 g/mol
MDL Number: MFCD06409221
CAS Number: 37904-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR