Anti-AQP3 Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-1253R-CY5.5
10281-078EA 823.27 USD
10281-078
Anti-AQP3 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Water channel required to promote glycerol permeability and water transport across cell membranes. Acts as a glycerol transporter in skin and plays an important role in regulating SC (stratum corneum) and epidermal glycerol content. Involved in skin hydration, wound healing, and tumorigenesis. Provides kidney medullary collecting duct with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient. Slightly permeable to urea and may function as a water and urea exit mechanism in antidiuresis in collecting duct cells. It may play an important role inMore Product Information