Differential Quick Stain, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26096-25 26096-50
102773-654EA 318.38 USD
102773-654 102773-656 102773-658
Differential Quick Stain, Electron Microscopy Sciences
Stains Hematology Stains
This kit is a Modified Giemsa Stain kit for quick turnaround results. It may be used for rapid blood smears for differential assessment, as well as for the detection of H. Pylori microorganisms.
  • The stain offers results in 15 seconds
  • Comes with a fixative for air dried cell suspensions and or touch preparation slides
Used for fine needle aspirations, frozen sections, blood smears, cytological specimens, cytopreps, microorganism detection and bone marrow biopsies.
Certificaciones: Certificates of analysis are available.
Información para pedidos: The Kit comes complete with 3 solutions and is available in 3 sizes.