Anti-TCP1A Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-12292R-CY3
10277-788EA 823.27 USD
10277-788
Anti-TCP1A Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Molecular chaperone; assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. Known to play a role, in vitro, in the folding of actin and tubulin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TCP1A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR