Anti-IGLL1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-16934
102708-820EA 635.51 USD
102708-820
Anti-IGLL1 Rabbit Polyclonal Antibody
Antibodies
The Lambda5 / IGLL1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Lambda5 / IGLL1. This antibody reacts with human. The Lambda5 / IGLL1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: IGLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol IGLL1
Antigen Synonyms AGM2|Immunoglobulin omega polypeptide|CD179b antigen|Immunoglobulin-related protein 14.1|VPREB2|IGL1|Ig lambda-5|IGO|CD179b|IGLJ14.1|CD179 antigen-like family member B|lambda5|IGL5CD179B|immunoglobulin lambda-like polypeptide 1|IGVPB|Pre-B lymphocyte-specific protein-2|immunoglobulin omega polypeptide chain|14.1|immunoglobulin-related 14.1 protein|IGLL
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 3543
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human IGLL1. The exact sequence is proprietary.
Purification Immunogen affinity purified
Concentration 0.4 mg/ml
Storage Buffer 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
Storage Temperature Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR