Ethyl-2-methyl-6-(4-nitrophenyl)nicotinate

Supplier: Matrix Scientific

080592-500MG
102670-302EA 193.85 USD
102670-302
Ethyl-2-methyl-6-(4-nitrophenyl)nicotinate
Ethyl-2-methyl-6-(4-nitrophenyl)nicotinate

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₄
MDL Number: MFCD08446619
CAS Number: 100970-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR