2-[(2-Furylmethyl)amino]-6-methyl-5-pentylpyrimidin-4(3H)-one

Supplier: Matrix Scientific

080512-500MG
102669-934EA 258.46 USD
102669-934
2-[(2-Furylmethyl)amino]-6-methyl-5-pentylpyrimidin-4(3H)-one
2-[(2-Furylmethyl)amino]-6-methyl-5-pentylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₁₅H₂₁N₃O₂
MDL Number: MFCD04196111
CAS Number: 669717-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR