N,N-Dimethylformamide 99,9 GC_ASSAY_METHOD

Supplier: CHEM IMPEX INTL INC MS
Danger

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

00575-18L 00575-1L 00575-1L
102570-980EA 855.12 USD
102570-980 102572-974 CH00575-1L
N,N-Dimethylformamide 99,9 GC_ASSAY_METHOD
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.944 g/mL
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
UN: 2265
ADR: 3 III
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless liquid
Color (APHA) ≤ 15
Water Content (Karl Fisher) ≤ 0.05%
Purity by GC ≥ 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR