Anti-HSPB2 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-11451R-CY5
10255-966EA 823.27 USD
10255-966
Anti-HSPB2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Advillin is an 819 amino acid protein that localizes to both the cytoplasm and the cytoskeleton and contains one HP domain and six gelsolin-like repeats. Expressed at high levels in colon and small intestine and at lower levels in uterus, thymus, testis and prostate, advillin functions as a calcium-regulated Actin-binding protein that may be involved in the development of neuronal cells, specifically those that form ganglia. The gene encoding advillin maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associatedMore Product Information