Phthalaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

074943-100G
102538-974EA 135.38 USD
102538-974
Phthalaldehyde ≥95%
Phthalaldehyde

Hazard: Toxic/Corrosive/Marine Pollutant/Refrigerate

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 83 °C (0.800 mmHg)
Melting Pt: 55…58 °C
Flash Pt: 132 °C
MDL Number: MFCD00003335
CAS Number: 643-79-8
UN: 2923
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR