3,5,6-Trichloropyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5,6-Trichloro-2-pyridinol

074357-5G
102536-004EA 220 USD
102536-004
3,5,6-Trichloropyridin-2(1H)-one ≥95%
3,5,6-Trichloropyridin-2(1H)-one

Hazard: Irritant

Formula: C₅H₂Cl₃NO
MW: 198.4345 g/mol
MDL Number: MFCD00130269
CAS Number: 6515-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR