4-Chlorosalicylaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-hydroxybenzaldehyde

072067-5G
102522-920EA 120 USD
102522-920
4-Chlorosalicylaldehyde ≥95%
4-Chlorosalicylaldehyde

Hazard: Irritant

Formula: C₇H₅ClO₂
MW: 156.5664 g/mol
MDL Number: MFCD06252499
CAS Number: 2420-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR