Human Recombinant Butyrophilin subfamily 3 member A2
Supplier: Bon Opus Biosciences
CC91-10UG CC91-1MG CC91-500UG CC91-50UG
102497-714EA 170.15 USD
102497-714 102497-716 102497-718 102497-720
Human Recombinant Butyrophilin subfamily 3 member A2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Butyrophilin 3A2 is produced by our Mammalian expression system and the target gene encoding Gln30-Trp248 is expressed with a 6His tag at the C-terminus.
Información accesorios: Accessories not included.
Información de suministro: Shipped Via FedEx
Envase: Shipped at ambient temperature via FedEx envelope.
Precaución: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.