LPS Extraction Kit, Bulldog Bio
Supplier: Bulldog Bio
17141
102408-112KT 216.92 USD
102408-112
LPS Extraction Kit, Bulldog Bio
Assays Cellular Assays
The LPS Extraction Kit provides a new approach to extracting LPS with higher yields, fewer steps and in far less time
  • Takes only 60 minutes to extract
  • Gives reproducibly high yields
  • Based on a robust water/phenol/chloroform extraction technology
Lipopolysaccharides (LPS) is a major constituent of the outer membrane of gram-negative bacteria. Some of LPS is a disease-causing endotoxin; it is also important in classification of serum type. Commonly using method is the complicated hot phenol-water extraction that takes long period of time.
Based on a robust water/phenol/chloroform extraction technology, the LPS extraction kit is flexible enough to run small culture volumes (<5 mL) or large (up to 100 mL) with similar yields.
Información de suministro: Kit includes 1x 100 mL lysis buffer, 1x 80 mL purification buffer.