Cooling Bath Circulators, CC-Range, Huber
Supplier: Huber
2011.0019.01
10220-780EA 57598.85 USD
10220-780 10220-782 10220-784 10220-786 10220-836 10220-838 10221-018 10221-022 10221-024 10221-030 10221-036 10221-042 10221-090 10221-092 10221-094 10221-096 10221-098 10221-100 10221-712 10221-714 10221-716 10221-718 10221-720 10221-722 10221-724 10221-726 10221-894 10221-896
Cooling Bath Circulators, CC-Range, Huber
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Refrigeration bath circulators are suitable for controlling the temperature of externally connected applications and thermoregulation of objects directly in the thermostat bath
The new multi-touch controller Pilot ONE offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu in 11 languages. All important operating parameters and temperature values are neatly displayed on the touchscreen. Thanks to the new favourites menu, the one-click operation and the integrated technical glossary the operation is very easy, just like on a smartphone. Integrated USBMore Product Information