Sampler, MiniSampler, Bürkle
Supplier: BURKLE INC
5305-0010 5305-0011 5305-0100 5305-0110 5305-0209 5305-1020 6205-0180
10217-032EA 213.54 USD
10217-032 10217-034 10217-036 10217-038 10217-040 10217-044 76047-098 76047-100
Sampler, MiniSampler, Bürkle
Samplers Fluid Samplers
The single-use, flexible, suction hose can be quickly replaced, thus ensuring that the samples are never contaminated
If necessary, a new hose can be used for every single sample. Due to its small diameter (8mm) and flexibility, the hose can reach sampling areas which are otherwise inaccessible. Maximum suction lift is five meters, depending on viscosity.
Información accesorios: Accessory sample containers include a screw lid, and are ideal for sampling and transport. Made of PFA for super pure collection of samples, containers are autoclavable.
Información de suministro: SuppliedMore Product Information