Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB100-64714
102114-060EA 975.2 USD
102114-060
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (Biotin)
Antibodies
Rabbit Polyclonal Mycoplasma pneumoniae Antibody [Biotin]. Tested Applications: ELISA, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Bacteria, Bovine (-), Human (-).

Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR