MegaMag Genie™ Magnetic Stirrers, 230 V
Supplier: Electron Microscopy Sciences
72404-20 72404-AU 72404-BR 72404-EU
102098-030EA 967.04 USD
102098-030 102098-032 102098-034 102098-036
MegaMag Genie™ Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
The revolutionary single-position MagStir Genie provides low to high speed stirring and editable memory profiles
  • 230 V model
MagStir Genie may be used with any regular flask/beaker or spinner flask and a maximum sample weight of 5.0 kg. Speed range of 10 to 1800 rpm including reverse and interval stirring for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers. Three power levels are available for various sample viscosities. Low-profile magnetic stirrer uses microprocessor control for precise andMore Product Information