SUPER-NUOVA® Digital Multi-Position Hotplate/Stirrer, 220 V
Supplier: Electron Microscopy Sciences
72356-20
102097-952EA 4815.06 USD
102097-952
SUPER-NUOVA® Digital Multi-Position Hotplate/Stirrer, 220 V
Stirrers Magnetic Hotplate Stirrers
Product Description and Operation:
  • 220 V model
Multi-Position Stirrer and Stirring Hot Plates feature four in one performance, all of the features of the Super-Nuova with 4-position stirring ability. Precision control, 1 rpm stir control and 1°C temperature control. Smart performance with walk away timer, RS232 output, and user calibration function. Simple operation with a single knob controls both heating and stirring. Built-in Safety Features – Hot top warning system and user defined over temperature protection. Units are offer with seamless, easy-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable